missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific™

$135.54 - $135.54

Chemical Identifiers

CAS 79917-90-1
Molecular Formula C8H15ClN2
Molecular Weight (g/mol) 174.67
MDL Number MFCD03095425
InChI Key FHDQNOXQSTVAIC-UHFFFAOYSA-M
Synonym 1-butyl-3-methylimidazolium chloride, 1-n-butyl-3-methylimidazolium chloride, 3-butyl-1-methyl-1h-imidazol-3-ium chloride, bmim cl, bmimcl, c4mim chloride, dsstox_cid_11461, dsstox_gsid_31461, dsstox_rid_78878
PubChem CID 2734161
IUPAC Name 1-butyl-3-methylimidazol-3-ium;chloride
SMILES CCCCN1C=C[N+](=C1)C.[Cl-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC354090250
SDS View Documents Promotion Details
Thermo Fisher Scientific
354090250
25g
Each for $135.54
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Ionic liquid
Specifications

Chemical Identifiers

79917-90-1
174.67
FHDQNOXQSTVAIC-UHFFFAOYSA-M
2734161
CCCCN1C=C[N+](=C1)C.[Cl-]
C8H15ClN2
MFCD03095425
1-butyl-3-methylimidazolium chloride, 1-n-butyl-3-methylimidazolium chloride, 3-butyl-1-methyl-1h-imidazol-3-ium chloride, bmim cl, bmimcl, c4mim chloride, dsstox_cid_11461, dsstox_gsid_31461, dsstox_rid_78878
1-butyl-3-methylimidazol-3-ium;chloride

Specifications

79917-90-1
100.0
73.0°C
C8H15ClN2
MFCD03095425
15, 1584
1-butyl-3-methylimidazolium chloride, 1-n-butyl-3-methylimidazolium chloride, 3-butyl-1-methyl-1h-imidazol-3-ium chloride, bmim cl, bmimcl, c4mim chloride, dsstox_cid_11461, dsstox_gsid_31461, dsstox_rid_78878
FHDQNOXQSTVAIC-UHFFFAOYSA-M
1-butyl-3-methylimidazol-3-ium;chloride
2734161
Mass
170.1 mPa.s (80°C)
1-Butyl-3-methylimidazolium chloride
98.0
Cream to Yellow
98% min. (HPLC)
Authentic
Glass bottle
25g
Solubility in water: soluble
CCCCN1C=C[N+](=C1)C.[Cl-]
174.67
174.67
98+%
1% Max. (K.F.)
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if swallowed.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep a

GHS Signal Word: Danger

TSCA : Not on TSCA inventory: for R and D use only; not for manufacturing or commercial purposes.

SDS
Product Certifications

RUO – Research Use Only