missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Butyl-2-pyrrolidone, TCI America™

$103.89 - $282.64

Chemical Identifiers

CAS 3470-98-2
Molecular Formula C8H15NO
Molecular Weight (g/mol) 141.214
MDL Number MFCD00020876
InChI Key BNXZHVUCNYMNOS-UHFFFAOYSA-N
Synonym N-Butyl-2-azacyclopentanone
PubChem CID 18984
IUPAC Name 1-butylpyrrolidin-2-one
SMILES CCCCN1CCCC1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B125425ML View Documents TCI America
B125425ML
25mL Each for $282.64
 
B12545ML View Documents TCI America
B12545ML
5mL Each for $103.89
 
Specifications

Chemical Identifiers

3470-98-2
141.214
BNXZHVUCNYMNOS-UHFFFAOYSA-N
18984
CCCCN1CCCC1=O
C8H15NO
MFCD00020876
N-Butyl-2-azacyclopentanone
1-butylpyrrolidin-2-one

Specifications

137°C
25mL
C8H15NO
N-Butyl-2-azacyclopentanone
CCCCN1CCCC1=O
141.214
141.21
≥98.0% (GC)
Yellow
3470-98-2
MFCD00020876
BNXZHVUCNYMNOS-UHFFFAOYSA-N
1-butylpyrrolidin-2-one
18984
Clear Liquid at 20°C
1-Butyl-2-pyrrolidone
Safety and Handling

Safety and Handling

RTECSNumber : UY5746000

TSCA : No