missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Butyl-2,3-dimethylimidazolium Tetrafluoroborate 98.0+%, TCI America™
$96.22 - $388.21
Chemical Identifiers
CAS | 402846-78-0 |
---|---|
Molecular Formula | C9H17BF4N2 |
Molecular Weight (g/mol) | 240.05 |
MDL Number | MFCD03427618 |
InChI Key | VCAIYEJBOWHUGP-UHFFFAOYSA-N |
Synonym | 1-butyl-2,3-dimethylimidazolium tetrafluoroborate, 1-n-butyl-2,3-dimethylimidazolium tetrafluoroborate, 1-butyl-2,3-dimethylimidazol-1-ium tetrafluoroborate, 3-butyl-1,2-dimethylimidazol-1-ium tetrafluoroborate, bdmim bf4, acmc-209jcq, dsstox_cid_27885, dsstox_rid_82638, dsstox_gsid_47909, ksc235m3b |
PubChem CID | 2734244 |
IUPAC Name | 1-butyl-2,3-dimethyl-1H-imidazol-3-ium; tetrafluoroboranuide |
SMILES | F[B-](F)(F)F.CCCCN1C=C[N+](C)=C1C |
Chemical Identifiers
402846-78-0 | |
240.05 | |
VCAIYEJBOWHUGP-UHFFFAOYSA-N | |
2734244 | |
F[B-](F)(F)F.CCCCN1C=C[N+](C)=C1C |
C9H17BF4N2 | |
MFCD03427618 | |
1-butyl-2,3-dimethylimidazolium tetrafluoroborate, 1-n-butyl-2,3-dimethylimidazolium tetrafluoroborate, 1-butyl-2,3-dimethylimidazol-1-ium tetrafluoroborate, 3-butyl-1,2-dimethylimidazol-1-ium tetrafluoroborate, bdmim bf4, acmc-209jcq, dsstox_cid_27885, dsstox_rid_82638, dsstox_gsid_47909, ksc235m3b | |
1-butyl-2,3-dimethyl-1H-imidazol-3-ium; tetrafluoroboranuide |
Specifications
402846-78-0 | |
C9H17BF4N2 | |
5 g | |
1-butyl-2,3-dimethylimidazolium tetrafluoroborate, 1-n-butyl-2,3-dimethylimidazolium tetrafluoroborate, 1-butyl-2,3-dimethylimidazol-1-ium tetrafluoroborate, 3-butyl-1,2-dimethylimidazol-1-ium tetrafluoroborate, bdmim bf4, acmc-209jcq, dsstox_cid_27885, dsstox_rid_82638, dsstox_gsid_47909, ksc235m3b | |
F[B-](F)(F)F.CCCCN1C=C[N+](C)=C1C | |
240.05 | |
240.05 | |
Crystalline Lumps |
White-Yellow | |
MFCD03427618 | |
1759 | |
VCAIYEJBOWHUGP-UHFFFAOYSA-N | |
1-butyl-2,3-dimethyl-1H-imidazol-3-ium; tetrafluoroboranuide | |
2734244 | |
≥98.0% (HPLC,N) | |
1-Butyl-2,3-dimethylimidazolium Tetrafluoroborate |
Safety and Handling
TSCA : No