missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Butyl-2,3-dimethylimidazolium chloride, 99%, Thermo Scientific Chemicals
$80.62 - $299.89
Chemical Identifiers
CAS | 98892-75-2 |
---|---|
Molecular Formula | C9H17ClN2 |
Molecular Weight (g/mol) | 188.699 |
MDL Number | MFCD03093291 |
InChI Key | HHHYPTORQNESCU-UHFFFAOYSA-M |
Synonym | 1-butyl-2,3-dimethylimidazolium chloride, 1-butyl-2,3-dimethyl-1h-imidazol-3-ium chloride, 1-butyl-2,3-dimethylimidazol-1-ium chloride, 3-butyl-1,2-dimethylimidazol-1-ium chloride, acmc-209sba, dsstox_cid_27882, dsstox_rid_82635, dsstox_gsid_47906, 1-butyl-2,3-dimethylimidazoliumchloride, 1-butyl-2,3-dime-thylimidazolium chloride |
PubChem CID | 2734167 |
IUPAC Name | 1-butyl-2,3-dimethylimidazol-3-ium;chloride |
SMILES | CCCCN1C=C[N+](=C1C)C.[Cl-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH2727006
|
Thermo Scientific Chemicals
H2727006 |
5 g |
N/A
|
|
|||||
AAH2727018
|
Thermo Scientific Chemicals
H2727018 |
50 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
98892-75-2 | |
188.699 | |
HHHYPTORQNESCU-UHFFFAOYSA-M | |
2734167 | |
CCCCN1C=C[N+](=C1C)C.[Cl-] |
C9H17ClN2 | |
MFCD03093291 | |
1-butyl-2,3-dimethylimidazolium chloride, 1-butyl-2,3-dimethyl-1h-imidazol-3-ium chloride, 1-butyl-2,3-dimethylimidazol-1-ium chloride, 3-butyl-1,2-dimethylimidazol-1-ium chloride, acmc-209sba, dsstox_cid_27882, dsstox_rid_82635, dsstox_gsid_47906, 1-butyl-2,3-dimethylimidazoliumchloride, 1-butyl-2,3-dime-thylimidazolium chloride | |
1-butyl-2,3-dimethylimidazol-3-ium;chloride |
Specifications
98892-75-2 | |
C9H17ClN2 | |
5 g | |
HHHYPTORQNESCU-UHFFFAOYSA-M | |
1-butyl-2,3-dimethylimidazol-3-ium;chloride | |
2734167 | |
99% |
89.5°C to 93.2°C | |
MFCD03093291 | |
1-butyl-2,3-dimethylimidazolium chloride, 1-butyl-2,3-dimethyl-1h-imidazol-3-ium chloride, 1-butyl-2,3-dimethylimidazol-1-ium chloride, 3-butyl-1,2-dimethylimidazol-1-ium chloride, acmc-209sba, dsstox_cid_27882, dsstox_rid_82635, dsstox_gsid_47906, 1-butyl-2,3-dimethylimidazoliumchloride, 1-butyl-2,3-dime-thylimidazolium chloride | |
CCCCN1C=C[N+](=C1C)C.[Cl-] | |
188.699 | |
188.69 | |
1-Butyl-2,3-dimethylimidazolium chloride |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only