missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Butyl-1-methylpyrrolidinium Dicyanamide, TCI America™

$192.50 - $641.69

Chemical Identifiers

CAS 370865-80-8
Molecular Formula C11H20N4
Molecular Weight (g/mol) 208.309
MDL Number MFCD08458923
InChI Key LBHLGZNUPKUZJC-UHFFFAOYSA-N
PubChem CID 16213711
IUPAC Name 1-butyl-1-methylpyrrolidin-1-ium;cyanoiminomethylideneazanide
SMILES CCCC[N+]1(CCCC1)C.C(=[N-])=NC#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B54535G View Documents TCI America
B54535G
5g Each for $192.50
 
B545325G View Documents TCI America
B545325G
25g Each for $641.69
 
Specifications

Chemical Identifiers

370865-80-8
208.309
LBHLGZNUPKUZJC-UHFFFAOYSA-N
1-butyl-1-methylpyrrolidin-1-ium;cyanoiminomethylideneazanide
C11H20N4
MFCD08458923
16213711
CCCC[N+]1(CCCC1)C.C(=[N-])=NC#N

Specifications

Yellow
370865-80-8
MFCD08458923
CCCC[N+]1(CCCC1)C.C(=[N-])=NC#N
208.309
208.31
≥98.0% (T)
5g
C11H20N4
LBHLGZNUPKUZJC-UHFFFAOYSA-N
1-butyl-1-methylpyrrolidin-1-ium;cyanoiminomethylideneazanide
16213711
Clear Liquid at 20°C
1-Butyl-1-methylpyrrolidinium Dicyanamide
Safety and Handling

Safety and Handling

TSCA : No