missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Butyl-1-methylpyrrolidinium Chloride, TCI America™

$99.31 - $296.39

Chemical Identifiers

CAS 479500-35-1
Molecular Formula C9H20ClN
Molecular Weight (g/mol) 177.716
MDL Number MFCD08458922
InChI Key BOOXKGZZTBKJFE-UHFFFAOYSA-M
PubChem CID 11769095
IUPAC Name 1-butyl-1-methylpyrrolidin-1-ium;chloride
SMILES CCCC[N+]1(CCCC1)C.[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B33585G View Documents TCI America
B33585G
5g Each for $99.31
 
B335825G View Documents TCI America
B335825G
25g Each for $296.39
 
Specifications

Chemical Identifiers

479500-35-1
177.716
BOOXKGZZTBKJFE-UHFFFAOYSA-M
1-butyl-1-methylpyrrolidin-1-ium;chloride
C9H20ClN
MFCD08458922
11769095
CCCC[N+]1(CCCC1)C.[Cl-]

Specifications

White-Yellow
479500-35-1
MFCD08458922
CCCC[N+]1(CCCC1)C.[Cl-]
177.716
177.72
≥98.0% (T)
5g
C9H20ClN
BOOXKGZZTBKJFE-UHFFFAOYSA-M
1-butyl-1-methylpyrrolidin-1-ium;chloride
11769095
Crystal-Powder at 20°C
1-Butyl-1-methylpyrrolidinium Chloride
Safety and Handling

Safety and Handling

TSCA : No