Please login to your online account to display your discounted pricing

1-Butoxy-4-eth-1-ynylbenzene, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB09896DA

 View more versions of this product

Catalog No. BTB09896DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 174.24
Quantity 1g
Assay 97%
CAS Max % 100.0
Molecular Formula C12H14O
CAS 79887-15-3
MDL Number MFCD00173878
CAS Min % 96.5
Packaging Amber Glass Bottle
Assay Percent Range 97%
Linear Formula CH3(CH2)3OC6H4Cidentical toCH
Chemical Name or Material 1-Butoxy-4-eth-1-ynylbenzene

Specifications

Chemical Name or Material 1-Butoxy-4-eth-1-ynylbenzene
CAS 79887-15-3
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H14O
Linear Formula CH3(CH2)3OC6H4Cidentical toCH
Formula Weight 174.24
MDL Number MFCD00173878