1-Butoxy-4-eth-1-ynylbenzene, 97%, Thermo Scientific™
Supplier:  Thermo Scientific Chemicals BTB09896EA
 View more versions of this product
Catalog No. BTB09896EA
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Specifications
Specifications
1-Butoxy-4-eth-1-ynylbenzene
96.5
97%
C12H14O
MFCD00173878
1-butoxy-4-eth-1-ynylbenzene, benzene,1-butoxy-4-ethynyl, 4-n-butoxyphenylacetylene, 4-butoxyphenylacetylene, 1-ethynyl-4-butoxybenzene, 1-butoxy-4-ethynyl-benzene
CCCCOC1=CC=C(C=C1)C#C
174.24
174.24
79887-15-3
100.0
Amber Glass Bottle
CH3(CH2)3OC6H4Cidentical toCH
10g
JMLXBCSKFMMFGF-UHFFFAOYSA-N
1-butoxy-4-ethynylbenzene
2775120
97%
Chemical Identifiers
79887-15-3
174.24
JMLXBCSKFMMFGF-UHFFFAOYSA-N
2775120
CCCCOC1=CC=C(C=C1)C#C
C12H14O
MFCD00173878
1-butoxy-4-eth-1-ynylbenzene, benzene,1-butoxy-4-ethynyl, 4-n-butoxyphenylacetylene, 4-butoxyphenylacetylene, 1-ethynyl-4-butoxybenzene, 1-butoxy-4-ethynyl-benzene
1-butoxy-4-ethynylbenzene
SDS Documents