Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Butenyl ethyl ether, cis + trans, 98%

Chemical Identifiers

Chemical Name or Material 1-Butenyl ethyl ether
CAS 929-05-5
Name Note Cis + Trans
Molecular Formula C6H12O
Linear Formula CH3CH2CH=CHOCH2CH3
Formula Weight 100.16
MDL Number MFCD03427069
UN Number UN3271
Synonym 1-Ethoxy-1-butene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2672006

  SDS
alfa aesar™
H2672006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2672014

  SDS
alfa aesar™
H2672014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2672022

  SDS
alfa aesar™
H2672022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 100.16
Chemical Name or Material 1-Butenyl ethyl ether
Name Note Cis + Trans
Assay 98%
Molecular Formula C6H12O
Linear Formula CH3CH2CH=CHOCH2CH3
MDL Number MFCD03427069
CAS 929-05-5
UN Number UN3271
Synonym 1-Ethoxy-1-butene
Health Hazard 1 Danger
Boiling Point 94° to 95°C
Health Hazard 2 GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Flash Point 6°C (43°F)
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.