missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Butanol, Ultrapure, Spectrophotometric Grade, 99.0+%

Manufacturer:  Alfa Aesar™ 32443K2

 View more versions of this product

Catalog No. AA32443K2



Specifications

Specifications

1-Butanol
Spectrophotometric grade
≥99.0%
CH3(CH2)3OH
UN1120
14,1540
CCCCO
74.123
CHEBI:28885
Liquid
Alcohol-like
35° (95°F)
Colorless
7
71-36-3
Ultrapure
C4H10O
MFCD00002964
969148
LRHPLDYGYMQRHN-UHFFFAOYSA-N
butan-1-ol
263
74.12
0.810g/mL
1.3990
117°C to 118°C
-90°C
1L

Chemical Identifiers

71-36-3
74.123
LRHPLDYGYMQRHN-UHFFFAOYSA-N
CHEBI:28885
CCCCO
C4H10O
MFCD00002964
263
butan-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
H318-H226-H302-H335-H336-H315
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes skin irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTANOLS

RTECSNumber : EO1400000

TSCA : TSCA

SDS Documents