missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Butanol, TCI America™

$21.39 - $33.61

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00002964
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym Butyl Alcohol
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B070425ML View Documents TCI America
B070425ML
25mL Each for $21.39
 
B0704500ML View Documents TCI America
B0704500ML
500mL Each for $33.61
 
Specifications

Chemical Identifiers

71-36-3
74.123
LRHPLDYGYMQRHN-UHFFFAOYSA-N
263
butan-1-ol
C4H10O
MFCD00002964
Butyl Alcohol
CHEBI:28885
CCCCO

Specifications

118°C
-90°C
71-36-3
MFCD00002964
Butyl Alcohol
CCCCO
74.123
CHEBI:28885
Clear Liquid at 20°C
1-Butanol
Colorless
25mL
C4H10O
1120
LRHPLDYGYMQRHN-UHFFFAOYSA-N
butan-1-ol
263
74.12
≥99.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3049

RTECSNumber : EO1400000

TSCA : Yes