missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Butanol, HPLC Grade, 99%, Alfa Aesar™

Filtered through 0.2μm filters

Manufacturer:  Alfa Aesar 22925K7

 View more versions of this product

Catalog No. AA22925K7



Specifications

Specifications

1-Butanol
HPLC
C4H10O
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
CCCCO
74.123
CHEBI:28885
Liquid
4L
71-36-3
99%
MFCD00002964
LRHPLDYGYMQRHN-UHFFFAOYSA-N
butan-1-ol
263
74.12
0.810

Chemical Identifiers

71-36-3
74.123
LRHPLDYGYMQRHN-UHFFFAOYSA-N
263
butan-1-ol
C4H10O
MFCD00002964
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
CHEBI:28885
CCCCO
Safety and Handling

Safety and Handling

EINECSNumber : 200-751-6

SDS Documents