Please login to your online account to display your discounted pricing

1-Butanol, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$143.63 - $1,501.29

Chemical Identifiers

Chemical Name or Material 1-Butanol
CAS 71-36-3
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula CH3CH2CH2CH2OH
Formula Weight 74.12
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002036

 
avantor performance
905403
Narrow Mouth Amber Glass Bottle 4L Each for $497.39

02002038

 
avantor performance
905401
Narrow Mouth Amber Glass Bottle 500mL Each for $143.63

02002037

 
avantor performance
905433
Narrow Mouth PVC Coated Amber Bottle 4L Each for $1,501.29
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 74.12
Molecular Formula CH3CH2CH2CH2OH
Chemical Name or Material 1-Butanol
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.81kg
Flash Point 37.00°C
Quantity 4L
CAS 71-36-3