Please login to your online account to display your discounted pricing

1-Butanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles, Alfa Aesar™

$39.39 - $39.39

Chemical Identifiers

Chemical Name or Material 1-Butanol
CAS 71-36-3
Name Note Anhydrous
Molecular Formula CH3(CH2)3OH
Formula Weight 74.12
MDL Number MFCD00002964
UN Number UN1120
Beilstein 969148
Merck Index 14,1540
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA41867AK

 
alfa aesar
41867AK
packaged under Argon in resealable ChemSeal™ bottles 250mL Each for $39.39
Description & Specifications

Specifications

Melting Point −90° to −89°C
Formula Weight 74.12
Quantity 250mL
Chemical Name or Material 1-Butanol
TSCA TSCA
Odor Alcohol-like
Packaging packaged under Argon in resealable ChemSeal™ bottles
Refractive Index 1.3990
Name Note Anhydrous
Flash Point 35°C (95°F)
Boiling Point 117° to 118°C
Physical Form Liquid
pH 7
Sensitivity Hygroscopic
Assay 99.9%
Molecular Formula CH3(CH2)3OH
MDL Number MFCD00002964
CAS 71-36-3
UN Number UN1120
Beilstein 969148
Merck Index 14,1540
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H226-H302-H335-H336-H315;
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes skin irritation.
Density 0.810
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTANOLS
RTECS Number EO1400000
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H226-H302-H335-H336-H315;
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes skin irritation.