missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Butanol, ACS, >99.4%, Alfa Aesar™

Maximum level of impurities: Color (APHA) 10, Evaporation residue 0.005%, Titratable acid 0.0008meq/g, Carbonyl compounds (as butyraldehyde) 0.01%, Butyl ether 0.2%, H2O 0.1%

Manufacturer:  Alfa Aesar 31068K7

 View more versions of this product

Catalog No. AA31068K7



Specifications

Specifications

Colorless liquid
340°C (707°F)
5.6 hPa (4mm Hg) at 20°C
4L
ACS
C4H10O
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
LRHPLDYGYMQRHN-UHFFFAOYSA-N
butan-1-ol
263
74.12
1-Butanol
Alcool-like
Colorless
71-36-3
>99.4%
MFCD00002964
1-Butanol
CCCCO
74.123
CHEBI:28885
Liquid

Chemical Identifiers

71-36-3
74.123
LRHPLDYGYMQRHN-UHFFFAOYSA-N
263
butan-1-ol
C4H10O
MFCD00002964
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
CHEBI:28885
CCCCO
Safety and Handling

Safety and Handling

H318-H315-H226-H302-H335-H336: Causes serious eye damage. Causes skin irritation. Flammable liquid and vapour. Harmful if swallowed. May cause respiratory irritation. May cause drowsiness or dizziness.

DOTInformation : DOT Class: 3, Packaging Group: III

EINECSNumber : 200-751-6

SDS Documents