missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Butanol, 99+%, for spectroscopy, ACROS Organics™

Manufacturer:  Acros Organics 167695000

 View more versions of this product

Catalog No. AC167695000


 

Description

Description

For UV/VIS spectroscopy
Specifications

Specifications

1-Butanol
71-36-3
99.0
For spectroscopy
99+%
CH3(CH2)3OH
1120
15, 1542
LRHPLDYGYMQRHN-UHFFFAOYSA-N
butan-1-ol
263
74.12
0.8100g/mL
1.3980 to 1.4000
Solubility in water: 80g/L (20°C). Other solubilities: miscible with ethanol, ether and many other, organic solvents
0.05% max. (Coulometric)
2.95 mPa.s (20°C)
Undesignated
500mL
0.097 max. at 238nm, (1 cm cell vs HPLC-grade water), (1 cm cell vs HPLC-grade water) at 0nm, 1 max. at 205nm, 0.4 max. at 220nm, 0.1 max. at 240nm, 0.04 max. at 260nm, 0.02 max. at 280nm, 0.01 max. 330nm, 0.602 max. at 210nm, 0.046 max. at 245nm, 0.009 max. at 270nm
(1 cm cell vs HPLC-grade water)
100.0
≥99%
C4H10O
MFCD00002964
01, 367
n-Butyl alcohol, n-Butanol, n-Butyl alcohol, n-Butanol
CCCCO
74.123
CHEBI:28885
Liquid
Glass bottle
0.0005% max.
0.81
35°C
117.6°C
-89.0°C

Chemical Identifiers

71-36-3
74.123
LRHPLDYGYMQRHN-UHFFFAOYSA-N
263
C4H10O
MFCD00002964
n-Butyl alcohol, n-Butanol, n-Butyl alcohol, n-Butanol
CHEBI:28885
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause drowsiness or dizziness.
Causes serious eye damage.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 200-751-6

RTECSNumber : EO1400000

TSCA : TSCA

SDS Documents