Please login to your online account to display your discounted pricing

1-Butanethiol, 98%, AcroSeal™, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Butanethiol
CAS 109-79-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H10S
Linear Formula CH3(CH2)3SH
Formula Weight 90.18
MDL Number MFCD00004905
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC430878000

  SDS
thermo fisher scientific
AC430878000
AcroSeal™ Glass Bottle 800mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC430871000

  SDS
thermo fisher scientific
AC430871000
AcroSeal™ Glass Bottle in Tin Can 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 90.18
Quantity 800mL
Color Clear colorless
Packaging AcroSeal™ Glass Bottle
Density 0.84g/mL
CAS Max % 100.0
Infrared Spectrum Authentic
Synonym n-Butyl mercaptan
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00004905
Molecular Formula C4H10S
Chemical Name or Material 1-Butanethiol
Assay Percent Range 98%
CAS Min % 97.5
CAS 109-79-5
Linear Formula CH3(CH2)3SH
Refractive Index 1.4420 to 1.4440 (20°C, 589nm)
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Highly flammable liquid and vapor.