missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Butanethiol, 98%, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC430878000

 View more versions of this product

Catalog No. AC430878000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
90.18
Clear colorless
0.84g/mL
Authentic
MFCD00004905
1-Butanethiol
97.5
CH3(CH2)3SH
Liquid
800mL
AcroSeal™ Glass Bottle
100.0
n-Butyl mercaptan
C4H10S
98%
109-79-5
1.4420 to 1.4440 (20°C, 589nm)

Chemical Identifiers

109-79-5
100.0
C4H10S
90.18
n-Butyl mercaptan
≥97.5%
97.5
98%
CH3(CH2)3SH
MFCD00004905
1-Butanethiol

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger