Learn More
1-Butanethiol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 150800500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Butanethiol | |
109-79-5 | |
100.0 | |
Colorless | |
98.0°C | |
Authentic | |
Glass bottle | |
1.4420 to 1.4440 | |
MFCD00004905 | |
01, 370 | |
0.84 | |
1-butanethiol, butanethiol, butyl mercaptan, n-butyl mercaptan, n-butanethiol, butylthiol, thiobutyl alcohol, n-butylmercaptan, 1-mercaptobutane, 1-butyl mercaptan | |
WQAQPCDUOCURKW-UHFFFAOYSA-N | |
butane-1-thiol | |
8012 | |
98% |
98% | |
97.5 | |
-116.0°C | |
0.8400g/mL | |
0.5°C | |
98% | |
C4H10S | |
CH3(CH2)3SH | |
50 mL | |
02,53; 04,64 | |
15, 1580 | |
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether | |
CCCCS | |
90.18 | |
90.18 | |
Liquid |
Chemical Identifiers
109-79-5 | |
90.18 | |
WQAQPCDUOCURKW-UHFFFAOYSA-N | |
8012 | |
CCCCS |
C4H10S | |
MFCD00004905 | |
1-butanethiol, butanethiol, butyl mercaptan, n-butyl mercaptan, n-butanethiol, butylthiol, thiobutyl alcohol, n-butylmercaptan, 1-mercaptobutane, 1-butyl mercaptan | |
butane-1-thiol |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Highly flammable liquid and vapor.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
EINECSNumber : 203-705-3
RTECSNumber : EK6300000
TSCA : TSCA
RUO – Research Use Only