missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Butanethiol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 043421AC

 View more versions of this product

Catalog No. AA43421AC



Description

Description

1-Butanethiol is used as a model gaseous analyte for fast-responding membrane-free amperometric gas sensor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Butanethiol
-116°C
0.842 g/mL
2°C (36°F)
C4H10S
CH3(CH2)3SH
25 mL
1730908
14,1577
Soluble in water (0.59g/L at 20°C).
CCCCS
90.184
90.19
Liquid
109-79-5
Colorless
98°C
Stench
1.4425
MFCD00004905
UN2347
Air Sensitive
1-butanethiol, butanethiol, butyl mercaptan, n-butyl mercaptan, n-butanethiol, butylthiol, thiobutyl alcohol, n-butylmercaptan, 1-mercaptobutane, 1-butyl mercaptan
WQAQPCDUOCURKW-UHFFFAOYSA-N
butane-1-thiol
8012
98%

Chemical Identifiers

109-79-5
90.184
WQAQPCDUOCURKW-UHFFFAOYSA-N
8012
CCCCS
C4H10S
MFCD00004905
1-butanethiol, butanethiol, butyl mercaptan, n-butyl mercaptan, n-butanethiol, butylthiol, thiobutyl alcohol, n-butylmercaptan, 1-mercaptobutane, 1-butyl mercaptan
butane-1-thiol
Safety and Handling

Safety and Handling

GHS H Statement
H225-H302-H332-H319-H335
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H225-H302+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BUTYL MERCAPTAN

EINECSNumber : 203-705-3

RTECSNumber : EK6300000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only