Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Butanethiol, 98%

Chemical Identifiers

Chemical Name or Material 1-Butanethiol
CAS 109-79-5
Molecular Formula C4H10S
Linear Formula CH3(CH2)3SH
Formula Weight 90.19
MDL Number MFCD00004905
UN Number UN2347
Beilstein 1730908
Merck Index 14,1577
Synonym n-Butyl mercaptan
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43421AE

  SDS
alfa aesar™
43421AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA43421AC

  SDS
alfa aesar™
43421AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA43421AP

  SDS
alfa aesar™
43421AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 90.19
Quantity 100mL
Health Hazard 1 Danger
Physical Form Liquid
CAS 109-79-5
Synonym n-Butyl mercaptan
Health Hazard 2 GHS H Statement
H225-H302-H332-H319-H335
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 1730908
Linear Formula CH3(CH2)3SH
Refractive Index 1.4425
Assay 98%
MDL Number MFCD00004905
Density 0.842g/mL
Sensitivity Air Sensitive
Chemical Name or Material 1-Butanethiol
Melting Point −116°C
Odor Stench
Merck Index 14,1577
Health Hazard 3 GHS P Statement
P210-P260-P280h-P305+P351+P338-P403
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store in a well-ventilated place.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BUTYL MERCAPTAN
RTECS Number EK6300000
Flash Point 2°C (36°F)
Color Colorless
UN Number UN2347
Boiling Point 98°C
TSCA TSCA
Molecular Formula C4H10S
Safety and Handling

Danger

GHS P Statement
P210-P260-P280h-P305+P351+P338-P403
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store in a well-ventilated place.

GHS H Statement
H225-H302-H332-H319-H335
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.