Learn More
1-Butanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 396641000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ion pair chromatographySpecifications
1-Butanesulfonic acid, sodium salt | |
2386-54-1 | |
0.02 max. (C=0.5 M in water) at 260nm, 0.04 max. (C=0.5 M in water) at 230nm, 0.06 max. (C=0.5 M in water) at 220nm, 0.1 max. (C=0.5 M in water) at 210nm | |
>300.0°C | |
1% max. (150°C) | |
99+% | |
C4H9NaO3S | |
MFCD00007540 | |
sodium 1-butanesulfonate, sodium butane-1-sulfonate, 1-butanesulfonic acid sodium salt, sodium butylsulfonate, 1-butanesulfonic acid, sodium salt, sodium butane-1-sulphonate, sodium butanesulfonate, ipc-alks-4, 1-butanesulfonic acid, sodium salt 1:1, acmc-1cdot | |
XQCHMGAOAWZUPI-UHFFFAOYSA-M | |
sodium;butane-1-sulfonate | |
4096517 | |
99+% | |
Crystalline Powder |
Anhydrous | |
99.0 | |
100.0 | |
White | |
Authentic | |
Glass bottle | |
CH3(CH2)3SO3Na | |
100 g | |
Solubility in water: soluble. | |
[Na+].CCCCS([O-])(=O)=O | |
160.16 | |
160.17 | |
Ion Pair Chromatography |
Chemical Identifiers
2386-54-1 | |
160.16 | |
XQCHMGAOAWZUPI-UHFFFAOYSA-M | |
4096517 | |
[Na+].CCCCS([O-])(=O)=O |
C4H9NaO3S | |
MFCD00007540 | |
sodium 1-butanesulfonate, sodium butane-1-sulfonate, 1-butanesulfonic acid sodium salt, sodium butylsulfonate, 1-butanesulfonic acid, sodium salt, sodium butane-1-sulphonate, sodium butanesulfonate, ipc-alks-4, 1-butanesulfonic acid, sodium salt 1:1, acmc-1cdot | |
sodium;butane-1-sulfonate |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and
GHS Signal Word: Warning
EINECSNumber : 219-201-1
RUO – Research Use Only