Learn More
1-Bromoundecane, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1433314
Description
1-Bromoundecane is used in the preparation of Grignard reagent by reacting with Mg in THF (tetrahydrofuran). It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromoundecane is used in the preparation of Grignard reagent by reacting with Mg in THF (tetrahydrofuran). It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
Specifications
1-Bromoundecane | |
-9°C | |
112°C to 114°C (10 mmHg) | |
C11H23Br | |
MFCD00000223 | |
1098390 | |
Insoluble in water. | |
CCCCCCCCCCCBr | |
235.209 | |
235.22 |
693-67-4 | |
1.052 | |
>110°C (230°F) | |
1.4563 | |
25 g | |
undecyl bromide, undecane, 1-bromo, hendecyl bromide, n-undecyl bromide, bromoundecane, 1-bromo-undecane, n-undecyl-1-bromide, labotest-bb ltbb001165, undecylbromide, 1-bromanylundecane | |
IKPSIIAXIDAQLG-UHFFFAOYSA-N | |
1-bromoundecane | |
12744 | |
98% |
Chemical Identifiers
693-67-4 | |
235.209 | |
IKPSIIAXIDAQLG-UHFFFAOYSA-N | |
12744 | |
CCCCCCCCCCCBr |
C11H23Br | |
MFCD00000223 | |
undecyl bromide, undecane, 1-bromo, hendecyl bromide, n-undecyl bromide, bromoundecane, 1-bromo-undecane, n-undecyl-1-bromide, labotest-bb ltbb001165, undecylbromide, 1-bromanylundecane | |
1-bromoundecane |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H332-H335
EINECSNumber : 211-757-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only