missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromotridecane, TCI America™

$165.00 - $527.09

Chemical Identifiers

CAS 765-09-3
Molecular Formula C13H27Br
Molecular Weight (g/mol) 263.263
MDL Number MFCD00000227
InChI Key BFDNZQUBFCYTIC-UHFFFAOYSA-N
Synonym Tridecyl Bromide
PubChem CID 13000
IUPAC Name 1-bromotridecane
SMILES CCCCCCCCCCCCCBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B09355ML View Documents TCI America
B09355ML
5mL Each for $165.00
 
B093525ML View Documents TCI America
B093525ML
25mL Each for $527.09
 
Specifications

Chemical Identifiers

765-09-3
263.263
BFDNZQUBFCYTIC-UHFFFAOYSA-N
13000
CCCCCCCCCCCCCBr
C13H27Br
MFCD00000227
Tridecyl Bromide
1-bromotridecane

Specifications

150°C
5mL
C13H27Br
Tridecyl Bromide
CCCCCCCCCCCCCBr
263.263
263.26
≥98.0% (GC)
Yellow
765-09-3
MFCD00000227
BFDNZQUBFCYTIC-UHFFFAOYSA-N
1-bromotridecane
13000
Clear Liquid at 20°C
1-Bromotridecane
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0067

RTECSNumber : YD3100000

TSCA : Yes