missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromotetradecane, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC187380025

 View more versions of this product

Catalog No. AC187380025

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

1-Bromotetradecane
112-71-0
Authentic
Glass bottle
0.9300g/mL
Liquid
Clear yellow
C14H29Br
277.28
Myristyl bromide
98%
97.5
100.0
1.4590 to 1.4610 (20°C, 589nm)
2.5kg
≥97.5%
98%
CH3(CH2)13Br
MFCD00000228

Chemical Identifiers

112-71-0
100.0
C14H29Br
277.28
Myristyl bromide
≥97.5%
97.5
98%
CH3(CH2)13Br
MFCD00000228
1-Bromotetradecane, 98%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 203-999-3

TSCA : TSCA