Please login to your online account to display your discounted pricing

1-Bromopropane, 99%, ACROS Organics™


Quantity 250mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  107312500

 View more versions of this product

Catalog No. AC107312500

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥98.5%
Quantity 250mL
Color Clear colorless to light yellow
Chemical Name or Material 1-Bromopropane
Storage Information Product darkens during storage
Assay Percent Range 99%
Molecular Formula C3H7Br
Linear Formula CH3CH2CH2Br
Formula Weight 122.99
MDL Number MFCD00000254
CAS 106-94-5
Synonym n-Propyl bromide
CAS Min % 98.5
Health Hazard 1 Danger
Infrared Spectrum Authentic
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Causes skin irritation.
May cause damage to organs through prolonged or repeated exposure.
May damage fertility. May damage the unborn child.
May cause respiratory irritation.
Highly flamma
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Refractive Index 1.4320 to 1.4360 (20°C, 589nm)

Specifications

Chemical Name or Material 1-Bromopropane
CAS 106-94-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C3H7Br
Linear Formula CH3CH2CH2Br
Formula Weight 122.99
MDL Number MFCD00000254
Synonym n-Propyl bromide
Accessories

Danger

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Causes skin irritation.
May cause damage to organs through prolonged or repeated exposure.
May damage fertility. May damage the unborn child.
May cause respiratory irritation.
Highly flamma

SDS & Safety Documents