missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromopropane, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A10461AK

 View more versions of this product

Catalog No. AAA10461AK



Description

Description

1-Bromopropane is used as a solvent for adhesives, degreasing plastics and optics. It is used in the production of asphalt, synthetic fiber and aerosols. It is employed for cleaning the metal surfaces, electronic circuit boards, and multi-layered OLEDs (Organic Light Emitting Diodes). It serves to replace perchloroethylene as a dry cleaning solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromopropane is used as a solvent for adhesives, degreasing plastics and optics. It is used in the production of asphalt, synthetic fiber and aerosols. It is employed for cleaning the metal surfaces, electronic circuit boards, and multi-layered OLEDs (Organic Light Emitting Diodes). It serves to replace perchloroethylene as a dry cleaning solvent.

Solubility
Miscible with ethanol, water and diethyl ether.

Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Specifications

1-Bromopropane
-110°C
70°C to 71°C
C3H7Br
CH3CH2CH2Br
250 mL
505936
14,7845
Miscible with ethanol,water and diethyl ether.
CCCBr
122.993
CHEBI:47105
99%
106-94-5
1.353
21°C (70°F)
1.434
MFCD00000254
UN2344
Light sensitive
n-propyl bromide, propyl bromide, propane, 1-bromo, propane, bromo, bromopropane, 1-bromo-propane, 1-propyl bromide, ccris 30, unii-y9746dne68, n-propylbromide
CYNYIHKIEHGYOZ-UHFFFAOYSA-N
1-bromopropane
7840
123

Chemical Identifiers

106-94-5
122.993
CYNYIHKIEHGYOZ-UHFFFAOYSA-N
7840
1-bromopropane
C3H7Br
MFCD00000254
n-propyl bromide, propyl bromide, propane, 1-bromo, propane, bromo, bromopropane, 1-bromo-propane, 1-propyl bromide, ccris 30, unii-y9746dne68, n-propylbromide
CHEBI:47105
CCCBr
Safety and Handling

Safety and Handling

GHS H Statement
H225-H360-H373-H315-H319-H335-H336
Highly flammable liquid and vapor.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335-H336-H350-H360FD-H373

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BROMOPROPANES

EINECSNumber : 203-445-0

RTECSNumber : TX4110000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only