missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromopinacolone, 97+%

Chemical Identifiers

CAS 5469-26-1
Molecular Formula C6H11BrO
Formula Weight 179.06
MDL Number MFCD00000206
UN Number UN2810
Beilstein 506485
Synonym 1-Bromo-3,3-dimethyl-2-butanone
Chemical Name or Material 1-Bromopinacolone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1065509 SDS Alfa Aesar™
A1065509
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAA1065518 SDS Alfa Aesar™
A1065518
50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAA1065530 SDS Alfa Aesar™
A1065530
250g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
1-Bromopinacolone is used as an iIntermediate for triazole compounds ( a five-membered ring structure compound contains three nitrogens and two carbons in the ring) which have biological activities such as antiviral, antibacterial, antifungal and antituberculous.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, bases, heat and light.
Description & Specifications

Chemical Identifiers

5469-26-1
179.06
UN2810
1-Bromo-3,3-dimethyl-2-butanone
C6H11BrO
MFCD00000206
506485
1-Bromopinacolone

Specifications

5469-26-1
189° to 192°C
75°C (167°F)
C6H11BrO
MFCD00000206
506485
1-Bromopinacolone
−10°C
10g
Pungent
179.06
UN2810
1-Bromo-3,3-dimethyl-2-butanone
≥97%

Safety and Handling

GHS H Statement
H331-H318-H227-H315-H335
Toxic if inhaled.
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

RTECSNumber : EL7010000

Recommended Storage : 2°C to 8°C