Please login to your online account to display your discounted pricing

1-Bromopinacolone, 93%, ACROS Organics™


Quantity 25mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC306410250

 View more versions of this product

Catalog No. AC306410250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Color Clear pale yellow to yellow-brownish
Assay ≥91%
Formula Weight 179.06
Physical Form Liquid
Quantity 25mL
Density 1.33g/mL
CAS Max % 100.0
CAS Min % 91.0
Refractive Index 1.4656 to 1.4676 (20°C, 589nm)
Chemical Name or Material 1-Bromopinacolone
Assay Percent Range 93%
CAS 5469-26-1
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
MDL Number MFCD00000206
Synonym 1-Bromo-3, 3-dimethyl-2-butanone
Packaging Glass bottle
Linear Formula (CH3)3CCOCHBr
Infrared Spectrum Authentic
Molecular Formula C6H11BrO

Specifications

Chemical Name or Material 1-Bromopinacolone
CAS 5469-26-1
CAS Min % 91.0
CAS Max % 100.0
Assay ≥91%
Assay Percent Range 93%
Molecular Formula C6H11BrO
Linear Formula (CH3)3CCOCHBr
Formula Weight 179.06
MDL Number MFCD00000206
Synonym 1-Bromo-3, 3-dimethyl-2-butanone
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents