Learn More
1-Bromopinacolone, 93%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 306410050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Bromopinacolone | |
91.0 | |
-10°C | |
1.3300g/mL | |
75°C | |
91% min. (GC) | |
C6H11BrO | |
(CH3)3CCOCH2Br | |
5 mL | |
1-bromopinacolone, bromopinacolone, 1-bromo-3,3-dimethyl-2-butanone, 1-bromopinacolin, bromopinacolin, bromomethyl tert-butyl ketone, pivaloylmethyl bromide, tert-butyl bromomethyl ketone, 2-butanone, 1-bromo-3,3-dimethyl, omega-brompinakolin german | |
CC(C)(C)C(=O)CBr | |
179.06 | |
179.06 | |
Liquid |
5469-26-1 | |
100.0 | |
Brown-Yellow to Yellow | |
189°C to 192°C | |
Authentic | |
Glass bottle | |
1.4656 to 1.4676 | |
MFCD00000206 | |
1.33 | |
SAIRZMWXVJEBMO-UHFFFAOYSA-N | |
1-bromo-3,3-dimethylbutan-2-one | |
21642 | |
93% |
Chemical Identifiers
5469-26-1 | |
179.06 | |
SAIRZMWXVJEBMO-UHFFFAOYSA-N | |
21642 | |
CC(C)(C)C(=O)CBr |
C6H11BrO | |
MFCD00000206 | |
1-bromopinacolone, bromopinacolone, 1-bromo-3,3-dimethyl-2-butanone, 1-bromopinacolin, bromopinacolin, bromomethyl tert-butyl ketone, pivaloylmethyl bromide, tert-butyl bromomethyl ketone, 2-butanone, 1-bromo-3,3-dimethyl, omega-brompinakolin german | |
1-bromo-3,3-dimethylbutan-2-one |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
EINECSNumber : 226-794-
RUO – Research Use Only