missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromoperfluoroheptane, 97%

$33.47 - $330.01

Chemical Identifiers

CAS 375-88-2
Molecular Formula C7BrF15
Molecular Weight (g/mol) 448.957
MDL Number MFCD00013570
InChI Key VPQQZKWYZYVTMU-UHFFFAOYSA-N
Synonym 1-bromoperfluoroheptane, perfluoroheptyl bromide, 1-bromopentadecafluoroheptane, perfluoro-n-heptyl bromide, pentadecafluoroheptyl bromide, heptane, 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoro, acmc-1ad35, 1-bromopentadecafluoroheptane; perfluoroheptyl bromide, 1-bromanyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecakis fluoranyl heptane
PubChem CID 67819
IUPAC Name 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoroheptane
SMILES C(C(C(C(F)(F)F)(F)F)(F)F)(C(C(C(F)(F)Br)(F)F)(F)F)(F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1015603 View Documents Alfa Aesar™
A1015603
1g Each for $33.47
 
AAA1015606 View Documents Alfa Aesar™
A1015606
5g Each for $88.22
 
AAA1015614 View Documents Alfa Aesar™
A1015614
25g Each for $330.01
 
Specifications

Chemical Identifiers

375-88-2
448.957
VPQQZKWYZYVTMU-UHFFFAOYSA-N
67819
C(C(C(C(F)(F)F)(F)F)(F)F)(C(C(C(F)(F)Br)(F)F)(F)F)(F)F
C7BrF15
MFCD00013570
1-bromoperfluoroheptane, perfluoroheptyl bromide, 1-bromopentadecafluoroheptane, perfluoro-n-heptyl bromide, pentadecafluoroheptyl bromide, heptane, 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoro, acmc-1ad35, 1-bromopentadecafluoroheptane; perfluoroheptyl bromide, 1-bromanyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecakis fluoranyl heptane
1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoroheptane

Specifications

117°C to 119°C
375-88-2
CF3(CF2)6Br
1716336
VPQQZKWYZYVTMU-UHFFFAOYSA-N
1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoroheptane
67819
97%
1g
C7BrF15
MFCD00013570
1-bromoperfluoroheptane, perfluoroheptyl bromide, 1-bromopentadecafluoroheptane, perfluoro-n-heptyl bromide, pentadecafluoroheptyl bromide, heptane, 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoro, acmc-1ad35, 1-bromopentadecafluoroheptane; perfluoroheptyl bromide, 1-bromanyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecakis fluoranyl heptane
C(C(C(C(F)(F)F)(F)F)(F)F)(C(C(C(F)(F)Br)(F)F)(F)F)(F)F
448.957
448.96
1-Bromoperfluoroheptane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280g-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

Warning

TSCA : TSCA