missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromopentane, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 152522500

 View more versions of this product

Catalog No. AC152522500



Description

Description

Specifications

Specifications

1-Bromopentane
31°C
Glass bottle
1.21
Colorless to Yellow
250mL
110-53-2
100.0
C5H11Br
MFCD00000267
15, 596
Solubility in water: practically insoluble. Other solubilities: soluble in alcohol,miscible with ether
CCCCCBr
151.05
151.05
98%
1.2100g/mL
Authentic
1.4430 to 1.446
130°C
-95°C
98%
97.5
97.5% min. (GC)
CH3(CH2)4Br
01, 131
1-bromo-pentane, 1-pentyl bromide, 1-pentylbromide, amyl bromide, bromopentane, n-amyl bromide, n-pentyl bromide, pentane, 1-bromo, pentyl bromide, unii-z2s4r599p0
YZWKKMVJZFACSU-UHFFFAOYSA-N
1-bromopentane
8057
Liquid

Chemical Identifiers

110-53-2
151.05
YZWKKMVJZFACSU-UHFFFAOYSA-N
8057
CCCCCBr
C5H11Br
MFCD00000267
1-bromo-pentane, 1-pentyl bromide, 1-pentylbromide, amyl bromide, bromopentane, n-amyl bromide, n-pentyl bromide, pentane, 1-bromo, pentyl bromide, unii-z2s4r599p0
1-bromopentane
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-776-0

RTECSNumber : RZ9770000

TSCA : TSCA

SDS Documents