Learn More
1-Bromooctadecane, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A155880E
Description
1-Bromooctadecane is used in the preparation of shortened single-walled carbon nanotubes (s-SWCNTs). It is utilized to prepare octadecane in the presence of sodium borohydride as a catalyst. It is involved as a raw material for the preparation of dimethyldistearylammonium bromide, which is a bentonite modifier.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromooctadecane is used in the preparation of shortened single-walled carbon nanotubes (s-SWCNTs). It is utilized to prepare octadecane in the presence of sodium borohydride as a catalyst. It is involved as a raw material for the preparation of dimethyldistearylammonium bromide, which is a bentonite modifier.
Solubility
Soluble in acetone and most common organic solvents. Insoluble in water.
Notes
Incompatible with strong oxidizing agents and strong bases. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
Specifications
1-Bromooctadecane | |
25°C to 30°C | |
150°C to 152°C (0.1 mmHg) | |
Odorless | |
CH3(CH2)17Br | |
2500 g | |
octadecyl bromide, stearyl bromide, octadecane, 1-bromo, n-octadecyl bromide, bromooctadecane, octadecylbromide, 1-bromoctadecane, n-octadecylbromide, 1-bromooctodecane, bromo-n-octadecane | |
WSULSMOGMLRGKU-UHFFFAOYSA-N | |
1-bromooctadecane | |
8218 | |
97% |
112-89-0 | |
0.976 | |
>110°C (230°F) | |
C18H37Br | |
MFCD00000231 | |
774145 | |
Soluble in acetone and most common organic solvents. Insoluble in water. | |
CCCCCCCCCCCCCCCCCCBr | |
333.40 | |
333.39 |
Chemical Identifiers
112-89-0 | |
333.40 | |
WSULSMOGMLRGKU-UHFFFAOYSA-N | |
8218 | |
CCCCCCCCCCCCCCCCCCBr |
C18H37Br | |
MFCD00000231 | |
octadecyl bromide, stearyl bromide, octadecane, 1-bromo, n-octadecyl bromide, bromooctadecane, octadecylbromide, 1-bromoctadecane, n-octadecylbromide, 1-bromooctodecane, bromo-n-octadecane | |
1-bromooctadecane |
Safety and Handling
EINECSNumber : 204-013-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only