missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromooctadecane, 96%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 190020025

 View more versions of this product

Catalog No. AC190020025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

1-Bromooctadecane
112-89-0
100.0
214.0°C to 216.0°C (12.0mmHg)
Authentic
Glass bottle
1.4620 to 1.4640
MFCD00000231
01,I,69
octadecyl bromide, stearyl bromide, octadecane, 1-bromo, n-octadecyl bromide, bromooctadecane, octadecylbromide, 1-bromoctadecane, n-octadecylbromide, 1-bromooctodecane, bromo-n-octadecane
WSULSMOGMLRGKU-UHFFFAOYSA-N
1-bromooctadecane
8218
96%
96%
95.0
0.9700g/mL
>110°C
95% min. (GC)
C18H37Br
CH3(CH2)17Br
2.5kg
0.97
Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
CCCCCCCCCCCCCCCCCCBr
333.40
333.38

Chemical Identifiers

112-89-0
333.40
WSULSMOGMLRGKU-UHFFFAOYSA-N
8218
CCCCCCCCCCCCCCCCCCBr
C18H37Br
MFCD00000231
octadecyl bromide, stearyl bromide, octadecane, 1-bromo, n-octadecyl bromide, bromooctadecane, octadecylbromide, 1-bromoctadecane, n-octadecylbromide, 1-bromooctodecane, bromo-n-octadecane
1-bromooctadecane
Safety and Handling

Safety and Handling

EINECSNumber : 204-013-4

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only