missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromononane, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC107205000

 View more versions of this product

Catalog No. AC107205000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
207.15
Clear colorless to golden
01, I, 63
96.0
CH3(CH2)8Br
1-Bromononane
97%
Glass bottle
Liquid
500g
1.08g/mL
100.0
1.4530 to 1.4550 (20°C, 589nm)
C9H19Br
693-58-3
MFCD00000278
Authentic

Chemical Identifiers

693-58-3
100.0
C9H19Br
207.15
1-Bromononane
96.0
97%
CH3(CH2)8Br
MFCD00000278
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.