missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

1-Bromonaphthalene (Reagent), Fisher Chemical
Manufacturer: Fisher Chemical N9500
Description
EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.
Specifications
1-Bromonaphthalene | |
Pass Test | |
1.470 to 1.490 (at 25°C) | |
Yellow | |
500mL | |
C10H7Br | |
.alpha.-bromonaphthalene, .alpha.-naphthyl bromide, 1-bromo-naphthalene, 1-naphthyl bromide, alpha-bromonaphthalene, alpha-naphthyl bromide, bromonaphthalene, bromonapthalene, naphthalene, 1-bromo, naphthalene, bromo | |
C1=CC=C2C(=C1)C=CC=C2Br | |
207.07 | |
207.08 | |
Reagent |
1.489g/cm³ | |
Glass Bottle | |
279°C | |
-1°C | |
90-11-9,580-13-2 | |
MFCD00003868 | |
DLKQHBOKULLWDQ-UHFFFAOYSA-N | |
1-bromonaphthalene | |
7001 | |
Liquid |
Chemical Identifiers
90-11-9 | |
207.07 | |
DLKQHBOKULLWDQ-UHFFFAOYSA-N | |
7001 | |
C1=CC=C2C(=C1)C=CC=C2Br |
C10H7Br | |
MFCD00003868 | |
.alpha.-bromonaphthalene, .alpha.-naphthyl bromide, 1-bromo-naphthalene, 1-naphthyl bromide, alpha-bromonaphthalene, alpha-naphthyl bromide, bromonaphthalene, bromonapthalene, naphthalene, 1-bromo, naphthalene, bromo | |
1-bromonaphthalene |
Safety and Handling
