Please login to your online account to display your discounted pricing

1-Bromonaphthalene, 96%, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: fisher scientific acros  107155000

 View more versions of this product

Catalog No. AC107155000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥95%
Formula Weight 207.07
Quantity 500mL
Color Clear yellow to yellow-brown
Beilstein 05, 547
Merck Index 15, 1436
EINECS Number 201-965-2
TSCA TSCA
RTECS Number QJ1545000
CAS Min % 95.0
CAS Max % 100.0
Additional Information Vapor Pressure: 14.8hPa at 20°C
Chemical Name or Material 1-Bromonaphthalene
Refractive Index 1.6560 to 1.6590 (20°C, 589nm)
Name Note 96%
Assay Percent Range 96%
Molecular Formula C10H7Br
Linear Formula C10H7Br
MDL Number MFCD00003868
CAS 90-11-9
Health Hazard 1 Warning
Individual Impurity 2-bromonaphthalene: < 2.5%
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
CAS 580-13-2
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material 1-Bromonaphthalene
Name Note 96%
CAS 90-11-9
CAS Min % 95.0
CAS Max % 100.0
CAS 580-13-2
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C10H7Br
Linear Formula C10H7Br
Formula Weight 207.07
MDL Number MFCD00003868
Accessories

Warning

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents