Please login to your online account to display your discounted pricing

1-(Bromomethyl)adamantane, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB13836DA

 View more versions of this product

Catalog No. BTB13836DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 229.16
Quantity 1g
Assay 97%
Packaging Amber Glass Bottle
CAS 14651-42-4
Assay Percent Range 97%
Chemical Name or Material 1-(Bromomethyl)adamantane
Molecular Formula C11H17Br

Specifications

Chemical Name or Material 1-(Bromomethyl)adamantane
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H17Br
Formula Weight 229.16