Please login to your online account to display your discounted pricing

1-(Bromomethyl)adamantane, 97%, Maybridge

$393.88 - $393.88

Chemical Identifiers

Chemical Name or Material 1-(Bromomethyl)adamantane
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H17Br
Formula Weight 229.16
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BTB13836DA

  SDS
fisher scientific company
BTB13836DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BTB13836DE

  SDS
fisher scientific company
BTB13836DE
Amber Glass Bottle 5g Each for $393.88
Description & Specifications

Specifications

Formula Weight 229.16
Quantity 1g
Assay 97%
Packaging Amber Glass Bottle
CAS 14651-42-4
Assay Percent Range 97%
Chemical Name or Material 1-(Bromomethyl)adamantane
Molecular Formula C11H17Br