Please login to your online account to display your discounted pricing

1-(Bromomethyl)-3-phenoxybenzene, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-(Bromomethyl)-3-phenoxybenzene
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H11BrO
Formula Weight 263.13
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO07329CB

  SDS
fisher scientific company
MO07329CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 263.13
Quantity 250mg
Assay 97%
Molecular Formula C13H11BrO
Chemical Name or Material 1-(Bromomethyl)-3-phenoxybenzene
CAS 51632-16-7
Assay Percent Range 97%