Please login to your online account to display your discounted pricing

1-(Bromomethyl)-3-phenoxybenzene, 97%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  MO07329CB

 View more versions of this product

Catalog No. MO07329CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 250mg
Chemical Name or Material 1-(Bromomethyl)-3-phenoxybenzene
CAS 51632-16-7
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H11BrO
Formula Weight 263.13g/mol

Specifications

Chemical Name or Material 1-(Bromomethyl)-3-phenoxybenzene
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H11BrO
Formula Weight 263.13g/mol