Please login to your online account to display your discounted pricing

1-(Bromomethyl)-2-(phenoxymethyl)benzene, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC58508DA

 View more versions of this product

Catalog No. CC58508DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 277.16
Quantity 1g
Assay 97%
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C14H13BrO
CAS 861319-72-4
CAS Min % 96.5
Packaging Amber Glass Bottle
Chemical Name or Material 1-(Bromomethyl)-2-(phenoxymethyl)benzene
MDL Number MFCD08690311
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1-(Bromomethyl)-2-(phenoxymethyl)benzene
CAS 861319-72-4
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C14H13BrO
Formula Weight 277.16
MDL Number MFCD08690311
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.