Please login to your online account to display your discounted pricing

1-Bromoisoquinoline +95%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC392590010

 View more versions of this product

Catalog No. AC392590010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95.0%
Formula Weight 208.05
Physical Form Low Melting Solid
Quantity 1g
Color Light yellow to light brown
CAS Max % 100.0
CAS Min % 95.0
Molecular Formula C9H6BrN
MDL Number MFCD00234478
Chemical Name or Material 1-Bromoisoquinoline
CAS 1532-71-4
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 95+%
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS P Statement
Toxic if swallowed.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1-Bromoisoquinoline
CAS 1532-71-4
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95.0%
Assay Percent Range 95+%
Molecular Formula C9H6BrN
Formula Weight 208.05
MDL Number MFCD00234478
Accessories

GHS Signal Word: Warning

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Toxic if swallowed.
Causes serious eye damage.

SDS & Safety Documents