Please login to your online account to display your discounted pricing

1-Bromoisoquinolin-3-amine, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB09943DE

 View more versions of this product

Catalog No. BTB09943DE

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 223.07
Quantity 5g
Assay 97%
CAS Max % 100.0
Chemical Name or Material 1-Bromoisoquinolin-3-amine
Assay Percent Range 97%
Molecular Formula C9H7BrN2
MDL Number MFCD00102193
CAS 13130-79-5
CAS Min % 96.5
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 1-Bromoisoquinolin-3-amine
CAS 13130-79-5
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H7BrN2
Formula Weight 223.07
MDL Number MFCD00102193
Accessories

Warning

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.