missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromohexane, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 107045000

 View more versions of this product

Catalog No. AC107045000


 

Specifications Description & Specifications

Specifications

≥99.0%
Liquid
Clear
100.0
CH3(CH2)5Br
1.4465 to 1.4485 (20°C, 589nm)
C6H13Br
1-Bromohexane
0.05% max. (K.F.)
0.1% max. (as HBr)
165.07
500mL
99.0
hexanol (GC): < 0.5%
n-Hexyl bromide
111-25-1
99+%
Glass bottle
Authentic

Chemical Identifiers

111-25-1
100.0
C6H13Br
165.07
1-Bromohexane
99.0
99+%
CH3(CH2)5Br
n-Hexyl bromide
≥99.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning