missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromohexadecane, 98%

Manufacturer:  Alfa Aesar™ A1514730

 View more versions of this product

Catalog No. AAA1514730



Description

Description

Applications
1-Bromohexadecane is used in the preparation of soluble carbon nano-onions by covalent functionalization with hexadecyl chains, as extraction solvent in determination of endocrine-disrupting phenols (EDPs) in water samples by ultrasound-assisted emulsification microextraction method.

Solubility
Difficult to mix.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications

Specifications

1-Bromohexadecane
Odorless
16°C to 18°C
112-82-3
MFCD00000230
hexadecyl bromide, cetyl bromide, n-hexadecyl bromide, hexadecane, 1-bromo, 1-hexadecyl bromide, n-hexadecyl-1-bromide, bromohexadecane, unii-76gji7gvam, hexadecane, bromo, 76gji7gvam
CCCCCCCCCCCCCCCCBr
305.344
305.35
176°C (348°F)
336°C
250g
C16H33Br
773989
HNTGIJLWHDPAFN-UHFFFAOYSA-N
1-bromohexadecane
8213
98%

Chemical Identifiers

112-82-3
305.344
HNTGIJLWHDPAFN-UHFFFAOYSA-N
8213
CCCCCCCCCCCCCCCCBr
C16H33Br
MFCD00000230
hexadecyl bromide, cetyl bromide, n-hexadecyl bromide, hexadecane, 1-bromo, 1-hexadecyl bromide, n-hexadecyl-1-bromide, bromohexadecane, unii-76gji7gvam, hexadecane, bromo, 76gji7gvam
1-bromohexadecane
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents