missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromodecane, 98%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 163605000

 View more versions of this product

Catalog No. AC163605000



Specifications

Specifications

1-Bromodecane
94°C
Glass bottle
1.06
Colorless to Yellow
500g
112-29-8
100.0
C10H21Br
MFCD00000221
1-decyl bromide
MYMSJFSOOQERIO-UHFFFAOYSA-N
1-bromodecane
8173
Liquid
1.0600g/mL
Authentic
1.4550 to 1.457
238°C
-29.6°C
98%
97.5
97.5% (GC)
CH3(CH2)9Br
01, II, 130
Solubility in water: slightly soluble. Other solubilities: miscible with most common organic solvents
CCCCCCCCCCBr
221.18
221.18
98%

Chemical Identifiers

112-29-8
221.18
MYMSJFSOOQERIO-UHFFFAOYSA-N
8173
CCCCCCCCCCBr
C10H21Br
MFCD00000221
1-decyl bromide
1-bromodecane
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-955-3

RTECSNumber : HD6850000

TSCA : TSCA

SDS Documents