missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromoanthraquinone, TCI America™

$278.06 - $912.11

Chemical Identifiers

CAS 632-83-7
Molecular Formula C14H7BrO2
Molecular Weight (g/mol) 287.112
InChI Key CXTPIHZYOGDSLV-UHFFFAOYSA-N
PubChem CID 12444
IUPAC Name 1-bromoanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B26881G View Documents TCI America
B26881G
1g Each for $278.06
 
B26885G View Documents TCI America
B26885G
5g Each for $912.11
 
Specifications

Chemical Identifiers

632-83-7
287.112
12444
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)Br
C14H7BrO2
CXTPIHZYOGDSLV-UHFFFAOYSA-N
1-bromoanthracene-9,10-dione

Specifications

Undesignated
1g
C14H7BrO2
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)Br
287.112
287.11
≥95.0% (GC)
191°C
632-83-7
CXTPIHZYOGDSLV-UHFFFAOYSA-N
1-bromoanthracene-9,10-dione
12444
Crystal-Powder at 20°C
1-Bromoanthraquinone
Safety and Handling

Safety and Handling

TSCA : No