missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Bromoadamantane, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 149031000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1-Bromoadamantane | |
768-90-1 | |
100.0 | |
98.5% min. (GC) | |
C10H15Br | |
100g | |
Solubility in water: insoluble. Other solubilities: soluble in diethyl ether,chloroform,benzene,,methanol and ethanol | |
C1C2CC3CC1CC(C2)(C3)Br | |
215.13 | |
215.13 |
99% | |
98.5 | |
Authentic | |
Glass bottle | |
MFCD00074721 | |
1-adamantyl bromide, adamantyl bromide, bromoadamantane, 1-bromo-adamantane, adamantane, 1-bromo, 1-bromotricyclo 3.3.1.13,7 decane, tricyclo 3.3.1.13,7 decane, 1-bromo, tricyclo 3.3.1.1 3,7-decane, 1-bromo, labotest-bb lt00239617, 3r,5s,7s-1-bromoadamantane | |
VQHPRVYDKRESCL-UHFFFAOYSA-N | |
1-bromoadamantane | |
79106 | |
99% |
Chemical Identifiers
768-90-1 | |
215.13 | |
VQHPRVYDKRESCL-UHFFFAOYSA-N | |
79106 | |
C1C2CC3CC1CC(C2)(C3)Br |
C10H15Br | |
MFCD00074721 | |
1-adamantyl bromide, adamantyl bromide, bromoadamantane, 1-bromo-adamantane, adamantane, 1-bromo, 1-bromotricyclo 3.3.1.13,7 decane, tricyclo 3.3.1.13,7 decane, 1-bromo, tricyclo 3.3.1.1 3,7-decane, 1-bromo, labotest-bb lt00239617, 3r,5s,7s-1-bromoadamantane | |
1-bromoadamantane |
Safety and Handling
MOISTURE SENSITIVE
EINECSNumber : 212-200-7
TSCA : TSCA
RUO â Research Use Only