Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-6-phenylhexane, 97%


Quantity 250mg

Manufacturer: alfa aesar™  L14568MD

 View more versions of this product

Catalog No. AAL14568MD

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 241.18
Chemical Name or Material 1-Bromo-6-phenylhexane
Assay 97%
Molecular Formula C6H5(CH2)6Br
MDL Number MFCD00236022
CAS 27976-27-8
Beilstein 2207649
Synonym 6-Phenyl-n-hexyl bromide
Boiling Point 115° to 118°C (6mmHg)
Quantity 250mg

Specifications

Chemical Name or Material 1-Bromo-6-phenylhexane
CAS 27976-27-8
Molecular Formula C6H5(CH2)6Br
Formula Weight 241.18
MDL Number MFCD00236022
Beilstein 2207649
Synonym 6-Phenyl-n-hexyl bromide